missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) 2,4-pentanedionate hydrate, Thermo Scientific Chemicals

262.31 € - 1025.10 €

Chemical Identifiers

CAS 206996-61-4
Molecular Formula C15H21CeO6
Molecular Weight (g/mol) 437.44
MDL Number MFCD00150164
InChI Key LCRQZRNVLFDPAZ-UHFFFAOYSA-N
Synonym cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
PubChem CID 16212434
SMILES [Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347997
View Documents
Thermo Scientific Alfa Aesar
021116.22
100 g
262.31 €
100g
Please to purchase this item. Need a web account? Register with us today!
11337997
View Documents
Thermo Scientific Alfa Aesar
021116.36
500 g
1025.10 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206996-61-4
437.44
LCRQZRNVLFDPAZ-UHFFFAOYSA-N
16212434
C15H21CeO6
MFCD00150164
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

131°C to 132°C
Red to Yellow
C15H21CeO6
cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
LCRQZRNVLFDPAZ-UHFFFAOYSA-N
cerium;(Z)-4-hydroxypent-3-en-2-one;hydrate
16212434
Hygroscopic
206996-61-4
Powder
MFCD00150164
Insoluble in water.
[Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
437.44
437.44 (Anhydrous)
Cerium(III) 2,4-pentanedionate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 239-720-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.