missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) acetate hydrate, 99.995% (metals basis), Thermo Scientific Chemicals

64.92 € - 874.35 €

Chemical Identifiers

CAS 206996-60-3
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.25
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
IUPAC Name cerium(3+) triacetate
SMILES [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044433.09
10 g
64.92 €
10g
Please to purchase this item. Need a web account? Register with us today!
11440730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044433.18
50 g
235.17 €
50g
Please to purchase this item. Need a web account? Register with us today!
11450730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044433.30
250 g
874.35 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206996-60-3
317.25
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
[Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H9CeO6
MFCD00051060
cerium(3+) triacetate

Specifications

206996-60-3
Crystalline
Ce(C2H3O2)3·xH2O
Soluble in water( 3.5g/L).
[Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
317.25
99.995%
Cerium (III) acetate hydrate
White
C6H9CeO6
MFCD00051060
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
cerium(3+) triacetate
317.26 (Anhydrous)
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only