missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11371079
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Specialty chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-00-8
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.248
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
PubChem CID 10826
IUPAC Name cerium(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]

Specifications

Melting Point 308°C (decomposition)
Physical Form Crystalline Aggregates
Quantity 50 g
Solubility Information Soluble in water (265g/L) at 15°C.
Formula Weight 344.28 (317.26 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Cerium(III) acetate sesquihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'