Learn More
Cerium(III) bromide hydrate, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Orange |
Physical Form | Crystalline |
Quantity | 50 g |
Merck Index | 14,1995 |
Solubility Information | Soluble in water. |
Formula Weight | 379.83 (Anhydrous) |
Percent Purity | 99.998% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Cerium (III) bromide hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.