missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide hydrate, 99.998% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 14457-87-5
Molecular Formula Br3Ce
Molecular Weight (g/mol) 379.83
MDL Number MFCD00016004
InChI Key MOOUSOJAOQPDEH-UHFFFAOYSA-K
IUPAC Name cerium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Ce+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044432.09
10 g N/A
N/A
11420730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044432.18
50 g N/A
N/A
Description

Description

Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14457-87-5
379.83
MOOUSOJAOQPDEH-UHFFFAOYSA-K
[Br-].[Br-].[Br-].[Ce+3]
Br3Ce
MFCD00016004
cerium(3+) tribromide

Specifications

14457-87-5
Crystalline
MFCD00016004
Soluble in water.
[Br-].[Br-].[Br-].[Ce+3]
379.83
99.998%
(metals basis)
Orange
Br3Ce
14,1995
MOOUSOJAOQPDEH-UHFFFAOYSA-K
cerium(3+) tribromide
379.83 (Anhydrous)
Odorless
Cerium (III) bromide hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-447-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only