missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) chloride, anhydrous, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11307997

Cerium(III) chloride, anhydrous, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11307997
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11307997

Brand: Thermo Scientific Alfa Aesar 021112.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Incandescent Gas Mantles, Spectrography, Polymerization of Olefins

Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.466
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chlorideShow MoreShow Less
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Boiling Point 1727°C
Melting Point 848°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5%
Solubility Information Soluble in water,alcohol,acetone,acids. Soluble in water,ethanol and acetone.
Formula Weight 246.48
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 3.92 g/cm3 at 20°C
Chemical Name or Material Cerium(III) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY13K043 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number021112.30,021112.18,021112.09
Lot NumberZ06J021 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number021112.30,021112.18,021112.09
Lot NumberU31J045 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number021112.30,021112.18,021112.09
Lot NumberS20J051 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog Number021112.30,021112,021112.18,021112.09
Lot NumberN28I056 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number021112.30,021112.18,021112.09
compliance-icons compliance-icons
Product Identifier
  • Cerium(III) chloride anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cerium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) chloride, anhydrous, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.