missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 18618-55-8 |
---|---|
Molecular Formula | CeCl3·7H2O |
Molecular Weight (g/mol) | 372.58 |
MDL Number | MFCD00149634 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10688124
|
Thermo Scientific Acros
211820250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10082710
|
Thermo Scientific Acros
211821000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
10732831
|
Thermo Scientific Acros
211825000 |
500 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18618-55-8 | |
CeCl3·7H2O | |
372.58 | |
MFCD00149634 |
Specifications
848.0°C | |
Yellow | |
98.5% min. (Argentometry) | |
CeCl3·7H2O | |
01,121; 09,99; 15,72; 16,4; 17,314 | |
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol | |
372.58 | |
3.97 |
18618-55-8 | |
Crystalline Powder and/or Crystals | |
CeCl3·7H2O | |
MFCD00149634 | |
10, 1956 | |
372.58 | |
99% | |
Cerium(III) chloride heptahydrate, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cerium(III) chloride heptahydrate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
RUO – Research Use Only