Learn More
Cerium(III) fluoride, 99.9% (REO), Thermo Scientific Chemicals
87.40 € - 1162.00 €
Chemical Identifiers
CAS | 7758-88-5 |
---|---|
Molecular Formula | CeF3 |
Molecular Weight (g/mol) | 197.111 |
MDL Number | MFCD00010932 |
InChI Key | QCCDYNYSHILRDG-UHFFFAOYSA-K |
Synonym | cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs |
PubChem CID | 24457 |
IUPAC Name | cerium(3+);trifluoride |
SMILES | [F-].[F-].[F-].[Ce+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377997
|
Thermo Scientific Alfa Aesar
021118.14 |
25 g |
87.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367997
|
Thermo Scientific Alfa Aesar
021118.22 |
100 g |
281.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357997
|
Thermo Scientific Alfa Aesar
021118.36 |
500 g |
1162.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(III) fluoride is used to make Lewis acid. It is considered to be the most effective glass polishing agent for high precision optical polishing. Cerium fluoride used as a core additive to improve the properties of the eventual weld. The fluoride provides stability to the arc and also modifies the behavior of the molten metal flowing into the weld.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-88-5 | |
197.111 | |
QCCDYNYSHILRDG-UHFFFAOYSA-K | |
24457 | |
[F-].[F-].[F-].[Ce+3] |
CeF3 | |
MFCD00010932 | |
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs | |
cerium(3+);trifluoride |
Specifications
∼2300°C | |
7758-88-5 | |
CeF3 | |
14,1998 | |
Practically insoluble in, but slowly hydrolyzed by water | |
[F-].[F-].[F-].[Ce+3] | |
197.111 | |
197.12 | |
Odorless | |
6.16 g/mL | |
Cerium(III) fluoride |
1460°C | |
Powder | |
MFCD00010932 | |
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs | |
QCCDYNYSHILRDG-UHFFFAOYSA-K | |
cerium(3+);trifluoride | |
24457 | |
99.9% | |
Hygroscopic | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
missing translation for 'einecsNumber' : 231-841-3
missing translation for 'rtecsNumber' : FK6125000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only