missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

For vacuum deposition

98.19 € - 294.47 €

Chemical Identifiers

CAS 7758-88-5
Molecular Formula CeF3
Molecular Weight (g/mol) 197.111
MDL Number MFCD00010932
InChI Key QCCDYNYSHILRDG-UHFFFAOYSA-K
Synonym cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
PubChem CID 24457
IUPAC Name cerium(3+);trifluoride
SMILES [F-].[F-].[F-].[Ce+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364108
View Documents
Thermo Scientific Alfa Aesar
041408.09
10 g
98.19 €
10g
Please to purchase this item. Need a web account? Register with us today!
11374108
View Documents
Thermo Scientific Alfa Aesar
041408.18
50 g
294.47 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-88-5
197.111
QCCDYNYSHILRDG-UHFFFAOYSA-K
24457
[F-].[F-].[F-].[Ce+3]
CeF3
MFCD00010932
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
cerium(3+);trifluoride

Specifications

∼2300°C
7758-88-5
CeF3
14,1998
QCCDYNYSHILRDG-UHFFFAOYSA-K
cerium(3+);trifluoride
24457
99.9%
Hygroscopic
(REO)
1460°C
Pieces
MFCD00010932
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs
[F-].[F-].[F-].[Ce+3]
197.111
197.12
Odorless
6.16 g/mL
Cerium(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

missing translation for 'einecsNumber' : 231-841-3

missing translation for 'rtecsNumber' : FK6125000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only