missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) nitrate hydrate, REacton™, 99.998% (metals basis)

Product Code. p-4407353
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11374868

Brand: Thermo Scientific Alfa Aesar 043181.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ce(NO3)3·xH2O (x ≈ 6-7)

Cerium(III) nitrate hydrate is used as an analytical reagent. It acts as a catalyst in petrochemical industry and in the pharmaceutical industry. Further, it is a chemical that helps burn tissue recover in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Ce(NO3)3·xH2O (x ≈ 6-7)
MDL Number MFCD00010926

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.998%
Solubility Information Soluble in water,alcohol and acetone
Formula Weight 326.13 (Anhydrous)
Odor Acidic
Sensitivity Hygroscopic
Appearance White crystalline
Density 1.67g/cm3 at 20°C
Chemical Name or Material Cerium(III) nitrate hydrate, 150°C - 3H2O
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.