missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) oxalate hydrate, 99%

Product Code. 11319387
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11319387

Brand: Thermo Scientific Alfa Aesar 040227.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15750-47-7 | C6Ce2O12 | 544.29 g/mol

It is used in ceramics, glass, phosphors and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15750-47-7
Molecular Formula C6Ce2O12
Molecular Weight (g/mol) 544.29
MDL Number MFCD00013082
InChI Key ZMZNLKYXLARXFY-UHFFFAOYSA-H
Synonym cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
PubChem CID 91998046
IUPAC Name cerium;oxalic acid;hydrate
SMILES [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
UN Number UN3288
Merck Index 14,2001
Solubility Information Insoluble in water; Soluble in hot dilute HCl and H2SO4
Formula Weight 544.29 (Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) oxalate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.