missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

32.76 € - 902.00 €

Chemical Identifiers

CAS 15750-47-7
Molecular Formula C6Ce2O12
Molecular Weight (g/mol) 544.29
MDL Number MFCD00013082
InChI Key ZMZNLKYXLARXFY-UHFFFAOYSA-H
Synonym cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
PubChem CID 91998046
SMILES [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338007
View Documents
Thermo Scientific Alfa Aesar
021122.06
5 g
32.76 €
5g
Please to purchase this item. Need a web account? Register with us today!
11328007
View Documents
Thermo Scientific Alfa Aesar
021122.14
25 g
85.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
11318007
View Documents
Thermo Scientific Alfa Aesar
021122.22
100 g
256.28 €
100g
Please to purchase this item. Need a web account? Register with us today!
11308007
View Documents
Thermo Scientific Alfa Aesar
021122.36
500 g
902.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15750-47-7
544.29
ZMZNLKYXLARXFY-UHFFFAOYSA-H
91998046
C6Ce2O12
MFCD00013082
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

(decomposition)
Powder
MFCD00013082
14,2001
ZMZNLKYXLARXFY-UHFFFAOYSA-H
cerium;oxalic acid;hydrate
91998046
99.9%
Odorless
(REO)
15750-47-7
C6Ce2O12
UN3288
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
544.29
544.29 (Anhydrous)
Reacton™
Hygroscopic
Cerium(III) oxalate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.