missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) phosphate, 99% min, Thermo Scientific Chemicals

52.80 € - 558.00 €

Chemical Identifiers

CAS 13454-71-2
Molecular Formula CeO4P
Molecular Weight (g/mol) 235.09
MDL Number MFCD00049424
InChI Key TYAVIWGEVOBWDZ-UHFFFAOYSA-K
Synonym cerium phosphate, cerium 3+ phosphate, unii-xr1z4biw2j, cerium iii phosphate, xr1z4biw2j, phosphoric acid,cerium 3+ salt 1:1, cerium monophosphate, cerium 3 phosphate, cerium orthophosphate cepo4, acmc-20alhk
PubChem CID 159673
IUPAC Name cerium(3+);phosphate
SMILES [Ce+3].[O-]P([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039215.14
25 g
52.80 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11373608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039215.22
100 g
150.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11363608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039215.36
500 g
558.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Intermediate product in cerium-based technique for staining lysosomal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13454-71-2
235.09
TYAVIWGEVOBWDZ-UHFFFAOYSA-K
159673
[Ce+3].[O-]P([O-])([O-])=O
CeO4P
MFCD00049424
cerium phosphate, cerium 3+ phosphate, unii-xr1z4biw2j, cerium iii phosphate, xr1z4biw2j, phosphoric acid,cerium 3+ salt 1:1, cerium monophosphate, cerium 3 phosphate, cerium orthophosphate cepo4, acmc-20alhk
cerium(3+);phosphate

Specifications

13454-71-2
Powder
CePO4
cerium phosphate, cerium 3+ phosphate, unii-xr1z4biw2j, cerium iii phosphate, xr1z4biw2j, phosphoric acid,cerium 3+ salt 1:1, cerium monophosphate, cerium 3 phosphate, cerium orthophosphate cepo4, acmc-20alhk
TYAVIWGEVOBWDZ-UHFFFAOYSA-K
cerium(3+);phosphate
159673
≤99%
5.22 g/mL
Yellow
CeO4P
MFCD00049424
Insoluble in water.
[Ce+3].[O-]P([O-])([O-])=O
235.09
235.09
Odorless
Cerium(III) phosphate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 236-637-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only