missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides)

Product Code. p-3212493 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10046182

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13454-94-9 | Ce2O12S3 | 568.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-94-9
Molecular Formula Ce2O12S3
Molecular Weight (g/mol) 568.43
MDL Number MFCD00010931
InChI Key OZECDDHOAMNMQI-UHFFFAOYSA-H
Synonym cerium sulfate, cerous sulfate, cerium iii sulfate, cerium sulphate, dicerium trisulfate, cerium sesquisulfate, unii-0vf70o14r1, sulfuric acid, cerium salt, dicerium trisulphate, sulphuric acid, cerium salt
PubChem CID 159674
IUPAC Name cerium(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 g
Assay Percent Range 46.0 to 50% (Ce) (Complexometry)
Linear Formula Ce2(SO4)3
Merck Index 15, 2001
Solubility Information Solubility in water: slightly soluble
Formula Weight 568.43
Percent Purity 99.90%
Packaging Glass bottle
Chemical Name or Material Cerium(III) sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Cerium(III) sulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Cerium(III) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.