missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

166.83 € - 630.14 €

Chemical Identifiers

CAS 10450-59-6
Molecular Formula Ce2O12S3·8H2O
Molecular Weight (g/mol) 712.55
MDL Number MFCD00149641
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10547563
SDS View Documents
Thermo Scientific Acros
192790500
50 g Glass bottle
166.83 €
50g
Please to purchase this item. Need a web account? Register with us today!
10017632
SDS View Documents
Thermo Scientific Acros
192792500
250 g Glass bottle
630.14 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

10450-59-6
Ce2O12S3·8H2O
712.55
MFCD00149641

Specifications

10450-59-6
Crystals and/or Chunks
Ce2O12S3·8H2O
MFCD00149641
10, 1961
712.55
99.99%
Cerium(III) sulfate octahydrate
White
38.0 to 40.5% (Ce)
Ce2(SO4)3·8H2O
09,99
Solubility in water: slightly soluble
712.55
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Cerium(III) sulfate octahydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.