missing translation for 'onlineSavingsMsg'
Learn More

Cerium ingot, 99% (REO), Thermo Scientific Chemicals

Product Code. 11315038
Click to view available options
Quantity:
250 g
1 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Gray
UN Number UN1333
Quantity 250 g
Percent Purity 99%
Odor Odorless
Assay (REO)
Chemical Name or Material Cerium ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'