Learn More
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min, Thermo Scientific Chemicals
Description
Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 130°C |
Color | Yellow |
Physical Form | Crystalline |
Quantity | 250 g |
Assay Percent Range | 94% min. |
Solubility Information | Soluble in water and insoluble in ethanol. |
Formula Weight | 632.56 (596.53 Anhydrous) |
Grade | ACS |
Appearance | Yellow crystalline |
Chemical Name or Material | Cerium(IV) ammonium sulfate dihydrate, For analysis ACS |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N20L031 | Certificate of Analysis | 24/02/2025 | 040194.A7,040194.A1,040194.30 |
Z17K002 | Certificate of Analysis | 23/01/2025 | 040194.A7,040194.A1,040194.30 |
U05K002 | Certificate of Analysis | 22/08/2024 | 040194.A7,040194.A1,040194.30 |
S25K029 | Certificate of Analysis | 19/07/2024 | 040194.A7,040194.A1,040194.30 |
S25K028 | Certificate of Analysis | 19/07/2024 | 040194.A7,040194.A1,040194.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.