missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min, Thermo Scientific Chemicals

Product Code. 11349337
Click to view available options
Quantity:
250 g
1 kg
5 kg

Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
Molecular Formula (NH4)4Ce(SO4)4·2H2O
MDL Number MFCD00149417
Synonym Ammonium cerium sulfate

Specifications

Melting Point 130°C
Color Yellow
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 94% min.
Solubility Information Soluble in water and insoluble in ethanol.
Formula Weight 632.56 (596.53 Anhydrous)
Grade ACS
Appearance Yellow crystalline
Chemical Name or Material Cerium(IV) ammonium sulfate dihydrate, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'