missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min

Product Code. p-4407355
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11329337

Brand: Thermo Scientific Alfa Aesar 040194.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O

Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
Molecular Formula (NH4)4Ce(SO4)4·2H2O
MDL Number MFCD00149417
Synonym Ammonium cerium sulfate

Specifications

Melting Point 130°C
Color Yellow
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 94% min.
Solubility Information Soluble in water and insoluble in ethanol.
Formula Weight 632.56 (596.53 Anhydrous)
Grade ACS
Appearance Yellow crystalline
Chemical Name or Material Cerium(IV) ammonium sulfate dihydrate, For analysis ACS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.