missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

59.70 € - 203.01 €

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11390468
View Documents
Thermo Scientific Alfa Aesar
043818.09
10 g
59.70 €
10g
Please to purchase this item. Need a web account? Register with us today!
11380468
View Documents
Thermo Scientific Alfa Aesar
043818.18
50 g
203.01 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium oxide is used as a raw material for the production of glass and ceramics as well as additive in paints and varnishes   It is also used in thermal spraying for welding and soldering products and for metal surface treatment products, in industrial polishing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O

Specifications

2600°C
Pieces
MFCD00010933
Insoluble in water.
O=[Ce]=O
172.114
CHEBI:79089
7.132 g/mL
1306-38-3
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
Cerium(IV) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-150-4

missing translation for 'rtecsNumber' : FK6310000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only