Learn More
Cerium(IV) sulfate, anhydrous, 97%, Thermo Scientific Chemicals
61.00 € - 759.00 €
Chemical Identifiers
CAS | 13590-82-4 |
---|---|
Molecular Formula | CeO8S2 |
Molecular Weight (g/mol) | 332.23 |
MDL Number | MFCD00148852 |
InChI Key | VZDYWEUILIUIDF-UHFFFAOYSA-J |
Synonym | ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate |
PubChem CID | 159684 |
IUPAC Name | cerium(4+);disulfate |
SMILES | [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Description
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.
Solubility
Soluble in water and dilute sulfuric acid.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.
Chemical Identifiers
13590-82-4 | |
332.23 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
159684 | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
CeO8S2 | |
MFCD00148852 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
cerium(4+);disulfate |
Specifications
13590-82-4 | |
MFCD00148852 | |
14,1990 | |
Soluble in water and dilute sulfuric acid. | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
332.23 | |
332.24 | |
Hygroscopic | |
Cerium(IV) sulfate, anhydrous |
CeO8S2 | |
UN3085 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
cerium(4+);disulfate | |
159684 | |
97% | |
3.91 |
Safety and Handling
GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 237-029-5
missing translation for 'rtecsNumber' : WS6960000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.