missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(IV) Sulfate Hydrate, Extra Pure, SLR, Low In Other Rare Earths, Fisher Chemical
Click to view available options
100g, Plastic powder jar
Specifications
Specifications
CAS | 10294-42-5 |
Color | Yellow |
pH | 1.6 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Molecular Formula | CeH9O12S2 |
MDL Number | 149427 |
InChI Key | ZBNJQJDKRZKDSV-UHFFFAOYSA-K |
SMILES | O.O.O.O.[Ce+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O |
IUPAC Name | cerium(3+) tetrahydrate hydrogen sulfate sulfate |
Show More |
Safety and Handling

Product Identifier
- Cerium(IV) sulfate tetrahydrate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction