Learn More
Cerium(IV) sulfate hydrate, REacton™, 99% (REO), Thermo Scientific Chemicals
180.90 € - 795.96 €
Chemical Identifiers
CAS | 95838-16-7 |
---|---|
Molecular Formula | CeO8S2 |
Molecular Weight (g/mol) | 332.23 |
MDL Number | MFCD00148852 |
InChI Key | VZDYWEUILIUIDF-UHFFFAOYSA-J |
Synonym | sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1 |
PubChem CID | 20223355 |
IUPAC Name | λ⁴-cerium(4+) hydrate disulfate |
SMILES | [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341079
|
Thermo Scientific Alfa Aesar
011323.22 |
100 g |
180.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331079
|
Thermo Scientific Alfa Aesar
011323.36 |
500 g |
795.96 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(IV) sulfate hydrate acts as oxidizing agent which is used for the generation of o-benzoquinone from catechol. It promotes the formation of radicals from dialkyl malonates, which undergo substitution reactions with furans and thiophenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95838-16-7 | |
332.23 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
20223355 | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
CeO8S2 | |
MFCD00148852 | |
sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1 | |
λ⁴-cerium(4+) hydrate disulfate |
Specifications
195°C (decomposition) | |
Crystalline | |
MFCD00148852 | |
sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
λ⁴-cerium(4+) hydrate disulfate | |
20223355 | |
99% | |
Moisture sensitive | |
(REO) |
95838-16-7 | |
CeO8S2 | |
UN3085 | |
Slightly soluble in water. Soluble in dil.sulfuric acid. | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
332.23 | |
332.24 (Anhydrous) | |
Reacton™ | |
3.01 g/mL | |
Cerium(IV) sulfate hydrate |
Safety and Handling
GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 237-029-5
missing translation for 'rtecsNumber' : WS6960000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only