missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate tetrahydrate, 98+%

Product Code. p-222798 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10276540

Brand: Thermo Scientific Acros 199131000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-42-5 | CeO8S2·4H2O | 404.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-42-5
Molecular Formula CeO8S2·4H2O
Molecular Weight (g/mol) 404.29
MDL Number MFCD00149427
InChI Key MEXSQFDSPVYJOM-UHFFFAOYSA-J
Synonym cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrate
PubChem CID 22723557
IUPAC Name cerium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]

Specifications

Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Ce(SO4)2·4H2O
Fieser 02,65; 15,73
Merck Index 14, 1990
Solubility Information Solubility in water: 38g/L (50°C). Other solubilities: soluble in dilute sulfuric acid,slowly soluble in cold,more readily soluble in,hot mineral acids
Formula Weight 404.29
Percent Purity ≥98%
Packaging Plastic bottle
Density 1.06
Chemical Name or Material Cerium(IV) sulfate tetrahydrate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Cerium(IV) sulfate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.