missing translation for 'onlineSavingsMsg'
Learn More

Cerium silicide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11318737
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12014-85-6
Molecular Formula CeSi2
Molecular Weight (g/mol) 196.286
MDL Number MFCD00168082
InChI Key BLHYVSKDMXEWHD-UHFFFAOYSA-N
Synonym cerium silicide, cerium disilicide
PubChem CID 6336874
IUPAC Name bis($l^{2}-silanylidene)cerium
SMILES [Si]=[Ce]=[Si]

Specifications

Melting Point 1620°C
Color Gray
Physical Form Pieces
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 196.29
Percent Purity 99.9%
Odor Odorless
Density 5.31 g/mL
Assay (REO)
Chemical Name or Material Cerium silicide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.