Learn More
Certified Reference Material Aniline Point, Jet Aviation Fuel, UKAS ISO 17034, (nominal value 58.48 °C), Paragon Scientific™
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Aniline Point ASTM D611
Brand:  Paragon Scientific CRMU-APKR
Product Code. 16010050
  • 230.00 € / 250mL
Explore more special offers
Description
Description
  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications
Specifications
250 mL
UN 1223
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Certified Reference Material Aniline Point, Jet Aviation Fuel, UKAS ISO 17034, (nominal value 58.48 °C), Paragon Scientific™ > Quantity: 250mL
Nominal values quoted. Certified Values will vary from batch to batch.