Learn More
Certified Reference Material Density Standard, Gasoline, UKAS ISO 17034 Certified 250mL (nominal value: 0.73632 g/mL at 15°C), Paragon Scientific™
Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.
Brand:  Paragon Scientific CRMU-DEGA
Product Code. 15431850
  • 230.00 € / 250mL
Explore more special offers
Description
Description
  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications
Specifications
250 mL
UN 1203
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Certified Reference Material Density Standard, Gasoline, UKAS ISO 17034 Certified 250mL (nominal value: 0.73632 g/mL at 15°C), Paragon Scientific™ > Quantity: 250mL
Nominal values quoted. Certified Values will vary from batch to batch.