Learn More
Cesium, 99.8% (metals basis), Thermo Scientific Chemicals
141.71 € - 1580.87 €
Chemical Identifiers
CAS | 7440-46-2 |
---|---|
Molecular Formula | Cs |
Molecular Weight (g/mol) | 132.91 |
MDL Number | MFCD00134037 |
InChI Key | TVFDJXOCXUVLDH-UHFFFAOYSA-N |
Synonym | caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium |
PubChem CID | 5354618 |
ChEBI | CHEBI:30514 |
SMILES | [Cs] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354648
|
Thermo Scientific Alfa Aesar
043005.03 |
1 g |
141.71 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344648
|
Thermo Scientific Alfa Aesar
043005.06 |
5 g |
321.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334648
|
Thermo Scientific Alfa Aesar
043005.18 |
50 g |
1580.87 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-46-2 | |
132.91 | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
5354618 | |
[Cs] |
Cs | |
MFCD00134037 | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
CHEBI:30514 |
Specifications
Silver | |
99.8% (Metals basis) | |
MFCD00134037 | |
TVFDJXOCXUVLDH-UHFFFAOYSA-N | |
caesium | |
5354618 | |
Cesium |
7440-46-2 | |
Cs | |
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium | |
[Cs] | |
132.91 | |
CHEBI:30514 |
Safety and Handling
P201-P202-P223-P231+P232-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H361
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I
missing translation for 'einecsNumber' : 231-155-4
missing translation for 'rtecsNumber' : FK9225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.