missing translation for 'onlineSavingsMsg'
Learn More

Cesium, 99.98% (metals basis), Thermo Scientific Chemicals

153.00 € - 2752.00 €

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010146.03
1 g
153.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11341198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010146.06
5 g
420.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11331198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010146.14
25 g
1257.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15490977
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010146.22
100 g
2752.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-46-2
132.91
TVFDJXOCXUVLDH-UHFFFAOYSA-N
5354618
[Cs]
Cs
MFCD00134037
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
CHEBI:30514

Specifications

Silver
Silver-colored solid
Cs
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
[Cs]
132.91
CHEBI:30514
Odorless
Cesium
7440-46-2
99.98% (Metals basis)
MFCD00134037
TVFDJXOCXUVLDH-UHFFFAOYSA-N
caesium
5354618
132.9054
0.0000011 hPa
Videos
Safety and Handling

Safety and Handling

P201-P202-P223-P231+P232-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H361

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I

missing translation for 'einecsNumber' : 231-155-4

missing translation for 'rtecsNumber' : FK9225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cesium

Signal Word
  • Danger

Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only