missing translation for 'onlineSavingsMsg'
Learn More
Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11322018

Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11322018
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11322018

Brand: Thermo Scientific Alfa Aesar 012929.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium Acetate is used in organic synthesis especially for Perkin reaction; formation of unsaturated cinnamic-type acids by the condensation of aromatic aldehydes with fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Physical Form Crystalline
Quantity 100 g
Solubility Information Soluble in water,DMSO.
Formula Weight 191.95
Percent Purity 99.9%
Odor Weak
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium acetate
compliance-icons
Product Identifier
  • Cesium acetate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, cesiumsalt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.