missing translation for 'onlineSavingsMsg'
Learn More

Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.96
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10186863
View Documents Promotion Details
Thermo Scientific Acros
390830250
25 g Glass bottle N/A
N/A
10433195
View Documents Promotion Details
Thermo Scientific Acros
390831000
100 g Glass bottle N/A
N/A
10196863
View Documents Promotion Details
Thermo Scientific Acros
390835000
500 g Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3396-11-0
191.96
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
5152919
CC(=O)[O-].[Cs+]
C2H3CsO2
MFCD00013056
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
cesium;acetate

Specifications

194.0°C
White
99%
CH3CO2Cs
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
cesium;acetate
5152919
99%
Authentic
3396-11-0
Crystalline Powder, Crystals and/or Chunks
C2H3CsO2
MFCD00013056
Solubility in water: 91.5% (21°C).
CC(=O)[O-].[Cs+]
191.96
191.96
Analytical
Cesium acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 222-248-

Product Identifier
  • Cesium acetate

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity Category 2

Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, cesiumsalt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only