missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium acetate, 99%, pure, Thermo Scientific Chemicals
64.82 € - 128.64 €
Chemical Identifiers
CAS | 3396-11-0 |
---|---|
Molecular Formula | C2H3CsO2 |
Molecular Weight (g/mol) | 191.96 |
MDL Number | MFCD00013056 |
InChI Key | ZOAIGCHJWKDIPJ-UHFFFAOYSA-M |
Synonym | cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion |
PubChem CID | 5152919 |
IUPAC Name | cesium;acetate |
SMILES | CC(=O)[O-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10256390
|
Thermo Scientific Acros
191040250 |
25 g | Glass bottle |
64.82 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10484145
|
Thermo Scientific Acros
191041000 |
100 g | Glass bottle |
128.64 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3396-11-0 | |
191.96 | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
5152919 | |
CC(=O)[O-].[Cs+] |
C2H3CsO2 | |
MFCD00013056 | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
cesium;acetate |
Specifications
3396-11-0 | |
White | |
C2H3CsO2 | |
MFCD00013056 | |
Solubility in water: 91.5% (21°C). | |
CC(=O)[O-].[Cs+] | |
191.96 | |
191.96 | |
Pure | |
Cesium acetate, Pure |
194.0°C | |
99% | |
CH3CO2Cs | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
cesium;acetate | |
5152919 | |
99% | |
Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 222-248-
RUO – Research Use Only