missing translation for 'onlineSavingsMsg'
Learn More

Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals

97.49 € - 361.80 €

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312018
SDS View Documents
Thermo Scientific Alfa Aesar
012928.18
50 g
97.49 €
50g
Please to purchase this item. Need a web account? Register with us today!
11302018
SDS View Documents
Thermo Scientific Alfa Aesar
012928.30
250 g
361.80 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cesium bromide is applied to make radiation detectors and other measuring devices. Cesium bromide has limited application in the deep IR. Cesium bromide is slightly more amenable to optical working than CsI and is sometimes utilized as a beam splitter component in wideband spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.