Learn More
Cesium carbonate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
For ultra trace analysis
40.30 € - 114.57 €
Chemical Identifiers
CAS | 534-17-8 |
---|---|
Molecular Formula | CCs2O3 |
Molecular Weight (g/mol) | 325.82 |
MDL Number | MFCD00010957 |
InChI Key | FJDQFPXHSGXQBY-UHFFFAOYSA-L |
Synonym | cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate |
PubChem CID | 10796 |
SMILES | [Cs+].[Cs+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10320744
|
Thermo Scientific Acros
278020100 |
10 g | Glass bottle |
40.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10206973
|
Thermo Scientific Acros
278020500 |
50 g | Glass bottle |
114.57 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
534-17-8 | |
325.82 | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
10796 |
CCs2O3 | |
MFCD00010957 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
[Cs+].[Cs+].[O-]C([O-])=O |
Specifications
>400.0°C | |
White | |
99.99% | |
Cs2CO3 | |
07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
dicaesium(1+) carbonate | |
10796 | |
99.99% | |
4.072 | |
Cesium carbonate |
534-17-8 | |
Powder | |
CCs2O3 | |
MFCD00010957 | |
15, 2009 | |
Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether | |
[Cs+].[Cs+].[O-]C([O-])=O | |
325.82 | |
325.82 | |
Trace Metal Basis | |
4.0720g/mL |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-591-9
- Cesium carbonate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P263-Avoid contact during pregnancy/while nursing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only