Learn More
Cesium carbonate, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 534-17-8 |
---|---|
Molecular Formula | CCs2O3 |
Molecular Weight (g/mol) | 325.82 |
MDL Number | MFCD00010957 |
InChI Key | FJDQFPXHSGXQBY-UHFFFAOYSA-L |
Synonym | cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate |
PubChem CID | 10796 |
SMILES | [Cs+].[Cs+].[O-]C([O-])=O |
Description
Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
534-17-8 | |
325.82 | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
10796 |
CCs2O3 | |
MFCD00010957 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
[Cs+].[Cs+].[O-]C([O-])=O |
Specifications
610°C (decomposition) | |
White | |
99.994% (Metal basis) | |
MFCD00010957 | |
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate | |
FJDQFPXHSGXQBY-UHFFFAOYSA-L | |
dicesium;carbonate | |
10796 | |
Odorless | |
White powder | |
Cesium carbonate |
534-17-8 | |
Powder/Granules | |
CCs2O3 | |
4546405 | |
Highly soluble in water. Soluble in alcohol,dimethyl sulfoxide and N,N-dimethylformamide. Insoluble in toluene and p-xylene. | |
[Cs+].[Cs+].[O-]C([O-])=O | |
325.82 | |
325.82 | |
Hygroscopic | |
4.072 |
Safety and Handling
P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
H318-H361f-H373
missing translation for 'einecsNumber' : 208-591-9
missing translation for 'rtecsNumber' : FK9400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only