missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11326717
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Analytical chemistry reagent for detecting inorganic ions. Source of cesium metal. Phase transfer catalyst in organic chemistry. Used for density gradient work to separate and purify proteins (isopycnic centrifugation) including chromosomal, plasmid and organelle (mitochondrial or plasmid) DNAs, and different RNA types. Preparation of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water and concentrated hydrochloric acid.
Formula Weight 168.36
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99
Chemical Name or Material Cesium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'