missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.999% (metals basis), Thermo Scientific Chemicals

90.15 € - 259.29 €

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391588
View Documents
Thermo Scientific Alfa Aesar
010993.14
25 g
90.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
11301598
View Documents
Thermo Scientific Alfa Aesar
010993.22
100 g
259.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds an application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrical conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
[Cl-].[Cs+]
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039

Specifications

1303°C
7647-17-8
Crystalline
ClCs
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
168.36
Hygroscopic
3.99
646°C
White
99.999% (Metal basis)
MFCD00010955
Soluble in water and concentrated hydrochloric acid.
[Cl-].[Cs+]
168.36
CHEBI:63039
1.6418
White powder/crystalline/beads
Cesium chloride
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H361-H373

missing translation for 'einecsNumber' : 231-600-2

missing translation for 'rtecsNumber' : FK9625000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cesium chloride

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity Category 2

Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Cesium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.