missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, MP Biomedicals™

Product Code. 11447891
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium Chloride, Ultra Pure Reagent, 99.999%, White Powder

  • Cesium chloride is used for the preparation of electrically conducting glasses.
  • It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Quantity 5 g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'