Learn More
Cesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
123.62 € - 1464.29 €
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
SMILES | [Cl-].[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330328
|
Thermo Scientific Alfa Aesar
043720.09 |
10 g |
123.62 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11310328
|
Thermo Scientific Alfa Aesar
043720.18 |
50 g |
393.96 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320328
|
Thermo Scientific Alfa Aesar
043720.30 |
250 g |
1464.29 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
[Cl-].[Cs+] |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 |
Specifications
1303°C | |
7647-17-8 | |
Beads | |
ClCs | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
24293 | |
168.36 | |
1.6418 | |
3.99 |
646°C | |
White | |
99.99% (Metals basis) | |
MFCD00010955 | |
Soluble in water and alcohol. Insoluble in acetone. | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
Odorless | |
Hygroscopic | |
Cesium chloride |
Safety and Handling
P201-P202-P260-P281-P308+P313-P501c
H361-H373
missing translation for 'einecsNumber' : 231-600-2
missing translation for 'rtecsNumber' : FK9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.