missing translation for 'onlineSavingsMsg'
Learn More
Cesium chloride, ultrapure, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15407149

Cesium chloride, ultrapure, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15407149
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15407149

Brand: Thermo Scientific Alfa Aesar J65950.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Color White
Physical Form Crystalline
Quantity 25 g
Merck Index 14,2011
Formula Weight 168.36
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99
Assay (metals basis)
Chemical Name or Material Cesium chloride
Grade Ultrapure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP11L009 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberJ65950.14,J65950.22
Lot NumberM04K007 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberJ65950.14,J65950.22
Lot NumberU15J032 Certificate TypeCertificate of Analysis Date21/08/2023 Catalog NumberJ65950.14,J65950.22
Lot NumberR12J010 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog NumberJ65950.14
Lot NumberN08J026 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ65950.14
compliance-icons
Product Identifier
  • Cesium chloride, ultrapure
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Cesium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium chloride, ultrapure, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.