missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents

Product Code. 10629743
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.999 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.999%
Grade DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Refractive Index 1.367 to 1.371 (at 25°C)
Chemical Name or Material Cesium Chloride
Show More Show Less
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'