Learn More
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals
45.20 € - 160.00 €
Chemical Identifiers
CAS | 13400-13-0 |
---|---|
Molecular Formula | CsF |
Molecular Weight (g/mol) | 151.90 |
MDL Number | MFCD00010960 |
InChI Key | XJHCXCQVJFPJIK-UHFFFAOYSA-M |
Synonym | cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride |
PubChem CID | 25953 |
SMILES | [F-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10392823
|
Thermo Scientific Acros
315910050 |
5 g | Plastic bottle |
45.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10790781
|
Thermo Scientific Acros
315910250 |
25 g | Plastic bottle |
104.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743144
|
Thermo Scientific Acros
315911000 |
100 g | Plastic bottle |
160.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
13400-13-0 | |
151.90 | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
25953 |
CsF | |
MFCD00010960 | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
[F-].[Cs+] |
Specifications
1251.0°C | |
13400-13-0 | |
Powder | |
CsF | |
MFCD00010960 | |
15, 2011 | |
Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine | |
[F-].[Cs+] | |
151.90 | |
151.91 | |
Extra Pure |
682.0°C | |
White | |
99.9% | |
CsF | |
01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68 | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
caesium(1+) fluoride | |
25953 | |
99.9% | |
Cesium fluoride |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
Contact with a
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISO
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 236-487-3
- Cesium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only