missing translation for 'onlineSavingsMsg'
Learn More
Cesium fluoride, 99%, for analysis, Thermo Scientific Chemicals Product Code.: 10781271

Cesium fluoride, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10781271
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10781271

Brand: Thermo Scientific Acros 189510250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251.0°C
Melting Point 682.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula CsF
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15,2011
Solubility Information Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
IUPAC Name caesium(1+) fluoride
Formula Weight 151.91
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Cesium fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium fluoride, 99%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.