missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water, Thermo Scientific Chemicals

Product Code. 10666982
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21351-79-1
Molecular Formula CsHO
Molecular Weight (g/mol) 149.91
MDL Number MFCD00010964
InChI Key HUCVOHYBFXVBRW-UHFFFAOYSA-M
Synonym cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
PubChem CID 62750
ChEBI CHEBI:33988
IUPAC Name cesium;hydroxide
SMILES [OH-].[Cs+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 g
Assay Percent Range 99.9%
Linear Formula CsOH
Merck Index 15, 2012
Solubility Information Solubility in water: soluble
Formula Weight 149.91
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 1.74
Density 1.7400g/mL
Chemical Name or Material Cesium hydroxide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.