Learn More
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water, Thermo Scientific Chemicals
129.00 € - 535.00 €
Chemical Identifiers
CAS | 21351-79-1 |
---|---|
Molecular Formula | CsHO |
Molecular Weight (g/mol) | 149.91 |
MDL Number | MFCD00010964 |
InChI Key | HUCVOHYBFXVBRW-UHFFFAOYSA-M |
Synonym | cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution |
PubChem CID | 62750 |
ChEBI | CHEBI:33988 |
IUPAC Name | cesium;hydroxide |
SMILES | [OH-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10793591
|
Thermo Scientific Acros
213601000 |
100 g | Plastic bottle |
129.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666982
|
Thermo Scientific Acros
213605000 |
500 g | Plastic bottle |
535.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
21351-79-1 | |
149.91 | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
62750 | |
cesium;hydroxide |
CsHO | |
MFCD00010964 | |
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution | |
CHEBI:33988 | |
[OH-].[Cs+] |
Specifications
21351-79-1 | |
Liquid | |
CsHO | |
MFCD00010964 | |
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
cesium;hydroxide | |
62750 | |
149.91 | |
Trace Metal Basis | |
1.7400g/mL |
Colorless | |
99.9% | |
CsOH | |
15, 2012 | |
Solubility in water: soluble | |
[OH-].[Cs+] | |
149.91 | |
CHEBI:33988 | |
99.9% | |
1.74 | |
Cesium hydroxide, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-344-1
missing translation for 'rtecsNumber' : FK9800000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only