missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | Soluble in water and ethanol. |
IUPAC Name | caesium(1+) hydroxide |
Formula Weight | 149.91 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction