Learn More
Cesium iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1280°C |
Melting Point | 626°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Merck Index | 14,2014 |
Solubility Information | Soluble in water. Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone. |
Formula Weight | 259.81 |
Percent Purity | 99.999% |
Odor | Odorless |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N07K006 | Certificate of Analysis | 13/02/2024 | 010992.22,010992.14 |
X10J041 | Certificate of Analysis | 17/10/2023 | 010992.22,010992.14 |
G25S031 | Certificate of Analysis | 08/12/2021 | 010992.22,010992.14 |
G18X063 | Certificate of Analysis | 07/12/2021 | 010992.22,010992.14 |
I02N46 | Certificate of Analysis | 07/12/2021 | 010992.22,010992.14 |
Safety and Handling


- Cesium iodide
- Warning
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- H400-Very toxic to aquatic life.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Cesium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.