missing translation for 'onlineSavingsMsg'
Learn More
Cesium methanesulfonate, 98%, Thermo Scientific Chemicals Product Code.: 11935891

Cesium methanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 11935891
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11935891

Brand: Thermo Scientific Acros 431580250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-61-0
Molecular Formula CH3CsO3S
Molecular Weight (g/mol) 228
InChI Key DJKJXRLREATOMF-UHFFFAOYSA-M
Synonym cesium methanesulfonate, csmes, csmsf, methanesulfonic acid cesium salt, caesium 1+ ion mesylate, cesium methanesulfonate used in patch clamp techniques, cesium methanesulfonate, used in patch clamp techniques
PubChem CID 5148066
IUPAC Name cesium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Cs+]

Specifications

Melting Point 262.0°C to 264.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Solubility Information (5% in water) Clear colorless
Formula Weight 228
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cesium methanesulfonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463879 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number431580050,431580250
Lot NumberA0461112 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number431580050,431580250
Lot NumberA0440626 Certificate TypeCertificate of Analysis Date31/03/2022 Catalog Number431580050,431580250
Lot NumberA0394978 Certificate TypeCertificate of Analysis Date14/09/2020 Catalog Number431580050,431580250
compliance-icons
Product Identifier
  • Cesium methanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium methanesulfonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.