missing translation for 'onlineSavingsMsg'
Learn More
Cesium nitrate, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11397607

Cesium nitrate, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11397607
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11397607

Brand: Thermo Scientific Alfa Aesar 012884.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of Cs compounds

Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Boiling Point (decomposition)
Melting Point 414°C
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in 5 parts cold and 0.5 parts boiling water. Soluble in acetone. Slightly soluble in alcoholSoluble in water and acetone. Slightly soluble in ethanol.
Formula Weight 194.91
Refractive Index 1.55
Sensitivity Hygroscopic
Density 3.68
Chemical Name or Material Cesium nitrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Caesium Nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cesium nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium nitrate, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.