Learn More
Cesium nitrate, 99.99% (metals basis), Thermo Scientific Chemicals
33.90 € - 1004.00 €
Chemical Identifiers
CAS | 7789-18-6 |
---|---|
Molecular Formula | CsNO3 |
Molecular Weight (g/mol) | 194.909 |
MDL Number | MFCD00010963 |
InChI Key | NLSCHDZTHVNDCP-UHFFFAOYSA-N |
Synonym | cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j |
PubChem CID | 62674 |
IUPAC Name | cesium;nitrate |
SMILES | [N+](=O)([O-])[O-].[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372388
|
Thermo Scientific Alfa Aesar
014440.06 |
5 g |
33.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392388
|
Thermo Scientific Alfa Aesar
014440.14 |
25 g |
95.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382388
|
Thermo Scientific Alfa Aesar
014440.22 |
100 g |
236.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302398
|
Thermo Scientific Alfa Aesar
014440.36 |
500 g |
1004.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-18-6 | |
194.909 | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
62674 | |
[N+](=O)([O-])[O-].[Cs+] |
CsNO3 | |
MFCD00010963 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
cesium;nitrate |
Specifications
(decomposition) | |
7789-18-6 | |
Crystalline | |
CsNO3 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
cesium;nitrate | |
62674 | |
Odorless | |
Hygroscopic | |
3.68 |
414°C | |
White | |
99.99% (Metal basis) | |
MFCD00010963 | |
Soluble in water and acetone. Slightly soluble in ethanol. | |
[N+](=O)([O-])[O-].[Cs+] | |
194.909 | |
194.91 | |
1.55 | |
White crystalline powder | |
Cesium nitrate |
Safety and Handling
P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c
H271-H302-H361f
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III
missing translation for 'einecsNumber' : 232-146-8
missing translation for 'rtecsNumber' : FL0700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only